Order Entry
United Kingdom
ContactUsLinkComponent
L(+)-Calcium lactate pentahydrate, Sigma-Aldrich®
L(+)-Calcium lactate pentahydrate, Sigma-Aldrich®
Catalog # SIAL21175-1KG-F
Supplier:  Merck
CAS Number:  
L(+)-Calcium lactate pentahydrate, Sigma-Aldrich®
Catalog # SIAL21175-1KG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :308.3 g/mol
Melting Pt:240 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:248-953-3
CAS Number:5743-47-5
Formula:C₆H₁₀CaO₆·5H₂O
MDL Number:MFCD00150344