Cerium(III) sulphate octahydrate, Sigma-Aldrich®
Catalog # SIAL203009-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:712.54
:M-Clarity™ quality level = MQ100
EINECS:236-644-6
CAS Number:10450-59-6
Formula:Ce₂O₁₂S₃·8H₂O
MDL Number:MFCD00149641