
Cerium(III) sulphate octahydrate ≥99.999% (metals basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10450-59-6
:
Formula:Ce₂O₁₂S₃·8H₂O
MDL Number:MFCD00149641
:712.54
Merck Index:14,2002
Density:2.886
EINECS:236-644-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together