(1S)-(-)-Borneol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:464-45-9
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00003759
Merck Index:13,01328
Boiling Pt:203 °C (1013 hPa)
Density:1.011 g/cm³ (20 °C)
ADR:4.1,III
:M-Clarity™ quality level = MQ100
:154.25 g/mol
EINECS:207-353-1
UN:1312
Frequently Bought Together