Disodium metasilicate nonahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium metasilicate nonahydrate
CAS Number:13517-24-3
:
Formula:Na₂SiO₃·9H₂O
:284.2 g/mol
Melting Pt:40 °C
Density:2.62 g/cm³ (21 °C)
EINECS:229-912-9
UN:3253
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together