Order Entry
United Kingdom
ContactUsLinkComponent
Hexachloroplatinic(IV) acid hydrate (∼40% Pt)
Corrosive
Toxic
Health
Environment
Hexachloroplatinic(IV) acid hydrate (∼40% Pt)
Certificates
undefined
Corrosive
Toxic
Health
Environment
Hexachloroplatinic(IV) acid hydrate (∼40% Pt)

Specification Test Results

  • Appearance:
    Orange to orange-brown Powder and/or chunks
  • Assay:
    37.5 to 40.5 % (Pt)
  • Total trace metal impurities:
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together