Order Entry
United Kingdom
ContactUsLinkComponent
3-Bromopropionic acid, Sigma-Aldrich®
Corrosive
3-Bromopropionic acid, Sigma-Aldrich®
Catalog # SIAL101281-100G
Supplier:  Merck
CAS Number:  
Corrosive
3-Bromopropionic acid, Sigma-Aldrich®
Catalog # SIAL101281-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.98 g/mol
Merck Index:13,01418
Boiling Pt:240 °C (1013 hPa)
Melting Pt:60 °C
Density:1.48 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-694-1
UN:3261
ADR:8,II
 :M-Clarity™ quality level = MQ100
 :GHS05
CAS Number:590-92-1
Formula:BrH₂CCH₂COOH
MDL Number:MFCD00002763