

Hydrochloric acid 36.5 - 38.0% ACS
Catalog # 33257.Q9
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:6
- Seq:0006
Specifications
About this item
:
Merck Index:14,4780
Boiling Pt:81.5…110 °C
Melting Pt:–30 °C
Density:1.19
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324