£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Gadolinium
Gadolinium ≥99.9% (by rare eath metals basis), ingot
Gadolinium ≥99.9% (by rare eath metals basis), ingot
45529.KF
:
Thermo Fisher Scientific
:
Gadolinium ≥99.9% (by rare eath metals basis), ingot
45529.KF
:
Thermo Fisher Scientific
:
Pk:
1
Seq:
0001
:
7440-54-2
:
:
Gd
:
MFCD00011022
:
157.25 g/mol
:
Packaged under argon.
:
Ambient
:
231-162-2