Order Entry
United Kingdom
ContactUsLinkComponent
 
Azomethine H sodium salt hydrate analytical reagent
  21645.124
 :  
undefined
Azomethine H sodium salt hydrate analytical reagent
  21645.124
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Suited for boron spectrophotometry:
    Passes test
  • Water:
    Max. 8 %
  • A (Lambda max.) (1 %; buffer pH 5.0):
    Min. 1100
  • Absorption maximum:
    231 - 241 nm

 

 

 :
 :463.42 g/mol
 :Ambient
 :227-698-1
 :206752-32-1
 :C₁₇H₁₂NNaO₈S₂·xH₂O
 :MFCD00149588