Resazurin sodium salt, tablets for milk analysis
330884Y
:
- Pack type:Aluminium tube
- Pk:100 Tab.
- Seq:0940
- Environmentally Preferable:
- Appearance:Red to purple tablets
- Shape:Round biconvex
- Average tablet weight:240 - 260 mg
- Diameter:7.50 - 8.00 mm
- Solubility in water (2 tablets/100 ml):Max. 4.0 min
: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin
:
:251.17 g/mol
:Ambient
:263-718-5
:62758-13-8
:C₁₂H₆NNaO₄
:MFCD00005036