Zinc chloride, special quality
Catalog # HONC72186-25KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Paper bag with inliner
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Assay:98.0 - 100.5 %
- Oxide chloride (as ZnO):complying
- pH (10 %, 20°C):4.6 - 5.3
- Ammonium (NH₄):Max. 0.03 %
- Arsenic (As):Max. 0.0002 %
- Calcium (Ca):Max. 0.01 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 0.02 %
- Magnesium (Mg):Max. 0.01 %
- Sodium (Na):Max. 0.01 %
- Lead (Pb):Max. 0.005 %
- Total N:Max. 0.005 %
- Sulfate (SO₄):Max. 200 ppm
- Al, Ca, Fe, Mg and heavy metals:complying
- Alkali and alkaline earth:Max. 1.0 %
- Residual solvents:complying
- Turbidity:Max. 3.5 NTU
Specification Test Results
About this item
:
:136.3 g/mol
Density:2.93 g/cm3
UN:2331
ADR:8,III
:GHS09
:GHS07
:GHS05
CAS Number:7646-85-7
Formula:ZnCl₂
MDL Number:MFCD00011295