Order Entry
United Kingdom
ContactUsLinkComponent
Sodium peroxodisulphate
Irritant
Health
Oxidizing
Sodium peroxodisulphate
Catalog # HONC13457-1KG
CAS Number:  
Irritant
Health
Oxidizing
Sodium peroxodisulphate
Catalog # HONC13457-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Iron (Fe):
    Max. 0.002 %
  • Heavy metals (as Pb):
    Max. 0.003 %
  • Chloride (Cl):
    Max. 0.005 %

Specification Test Results

About this item

 :
 :238.11 g/mol
UN:1505
ADR:5.1,III
 :GHS07
 :GHS08
 :GHS03
CAS Number:7775-27-1
Formula:Na₂S₂O₈
MDL Number:MFCD00003501