(+)-1-(9-Fluorenyl)ethyl chloroformate solution ≥18 mM in acetone, LiChropur™ for chiral derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:107474-79-3
:
Formula:C₁₆H₁₃ClO₂
Molecular Weight:272.73 g/mol
Storage Temperature:Refrigerator
UN:1090
ADR:3 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together