2′,3′-Dideoxycytidine 5′-triphosphate sodium salt ≥98.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
Catalog # 02241-1UMO-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1 µmol
- Seq:9050
Specifications
Specification Test Results
- Appearance (Color):Colorless
- Appearance (Form):Liquid
- Purity (HPLC area %):≥ 98.0 %
- HPLC minor component (area %):2',3'-DIDEOXYCYTIDINE 5'-DIPHOSPHATE ≤1.5 %
- Test by UV Spectroscopy:10.0 - 11.0 MMOL/L
- Extraneous activities:DNASES, RNASES NOT DETECTABLE
- A (260NM) /A (280NM):Report Result
- RECOMMENDED RETEST PERIOD:28 MONTHS
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ100
:451.16 g/mol
CAS Number:132619-66-0
Formula:C₉H₁₆N₃O₁₂P₃
MDL Number:MFCD00717536