Hexadecanamide ≥95%
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:629-54-9
:
Formula:C₁₆H₃₃NO
MDL Number:MFCD00025534
:255.45 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥95%
Specification Test Results
Frequently Bought Together