Order Entry
United Kingdom
ContactUsLinkComponent
 
 
 
 
Nitric acid ≥69%, Semiconductor grade (Honeywell 17952)
  HONC40222H-2.5L
 :  
 
 
 
Nitric acid ≥69%, Semiconductor grade (Honeywell 17952)
  HONC40222H-2.5L
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay:
    69 - 70 %
  • Residue on ignition (SO₄):
    Max. 5 ppm
  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.02 ppm
  • Boron (B):
    Max. 0.05 ppm
  • Barium (Ba):
    Max. 0.01 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.02 ppm
  • Calcium (Ca):
    Max. 0.1 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Gallium (Ga):
    Max. 0.02 ppm
  • Germanium (Ge):
    Max. 0.05 ppm
  • Indium (In):
    Max. 0.02 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.3 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.02 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.02 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Chloride (Cl):
    Max. 0.5 ppm
  • Phosphate (PO₄):
    Max. 0.5 ppm
  • Sulfate (SO₄):
    Max. 0.5 ppm

 

 

 :
 :63.01 g/mol
 :122 °C
 :–38 °C
 :1.420 g/cm3
 :Ambient
 :2031
 :8,II
 :GHS05
 :GHS06
 :GHS03
 :7697-37-2
 :HNO₃
 :MFCD00011349