Order Entry
United Kingdom
ContactUsLinkComponent
VWR®, Bottles, Rectangular, PET, Individually Wrapped
VWR®, Bottles, Rectangular, PET, Individually Wrapped
Catalog # 331-0290
Supplier:  VWR Collection
VWR®, Bottles, Rectangular, PET, Individually Wrapped
Catalog # 331-0290
Supplier:  VWR Collection
Specifications
  • Closure type:
    Screw cap
  • Version:
    Unlabelled, sodium thiosulphate dosed, sterile, individually wrapped
  • Capacity:
    500 ml
  • Seq:
    02
  • Cap colour:
    Red
  • Pk:
    120
Specifications
About this item
These bottles are made of PET and are perfect for sample collection, storage and transport.
  • Individually wrapped
  • Sodium thiosulfate dosed
  • Sterilised