Funnel, stainless steel
Supplier: BURKLE
Certificates
About this item
This funnel can be used to pour and transfer free-flowing powders, liquids and slightly viscous media easily.
- Made of stainless steel AISI 304 (1.4301)
- An air channel on the discharge tube ensures a complete, unobstructed flow
- Handle ensures easy handling
Depending on the diameter, this pouring aid is suitable for dispensing applications, from small amounts in laboratories to large quantities in industry.
The sturdy stainless-steel funnel can be used in a wide variety of applications.
Frequently Bought Together