Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphorhodamine G
Sulphorhodamine G fluorescent dye, for histology
Sulphorhodamine G fluorescent dye, for histology
Sulphorhodamine G fluorescent dye, for histology
Catalog # 80102.
Supplier:
Biotium
CAS Number:
Expand image
Sulphorhodamine G fluorescent dye, for histology
Catalog # 80102.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
CAS Number:
5873-16-5
MDL Number:
MFCD00213704
Product Details