Order Entry
United Kingdom
ContactUsLinkComponent
RuBP-2S fluorescent dye, for histology
RuBP-2S fluorescent dye, for histology
Catalog # 80024.
Supplier:  Biotium
CAS Number:  
undefined
RuBP-2S fluorescent dye, for histology
Catalog # 80024.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :1256.3 g/mol
Storage Temperature:Refrigerator
Formula:C₇₂H₄₆N₆O₆RuS₂