Supelco® Organic Reference Standard, Trans-4-Hydroxy-ʟ-Proline
Catalog # 41875-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:51-35-4
- Description:trans-4-Hydroxy-ʟ-proline
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00064320
- M-Clarity quality level:MQ100
- Synonym(s):(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid; Hyp
- Melting point:273 °C (dec.) (lit.)
- Formula:C₅H₉NO₃
- Molecular weight:131,13
- Storage temperature:2...8 °C
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.