Organic reference standard, Putrescine, analytical standard, Supelco®
51799-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:110-60-1
- Description:Putrescine, ≥98,5% (GC)
- Melting point:25...28 °C
- Formula:C₂H₁₂N₂
- Storage temperature:2...8 °C
- Description:Putrescine
- Boiling point:158...160 °C
- Form:Neat
Putrescine is a naturally occurring polyamine, which is produced via breakdown of ornithine amino acid.
Putrescine may be used as a standard in determining the concentration of putrescine present in fish and fishery products using thin layer chromatography (TLC) coupled with densitometry and high performance liquid chromatography (HPLC).