Order Entry
United Kingdom
ContactUsLinkComponent
Supelco® Organic Reference Standard, Oenothein B
Supelco® Organic Reference Standard, Oenothein B
Catalog # 03805-10MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Oenothein B
Catalog # 03805-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    104987-36-2
  • Description:
    Oenothein B
  • Environmentally Preferable:
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₆₈H₄₈O₄₄
  • Molecular weight:
    1569,08
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

About this item