Supelco® Organic Reference Standard, ʟ-Lysine
Catalog # 23128-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:56-87-1
- Description:ʟ-Lysine
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00064433
- M-Clarity quality level:MQ100
- Synonym(s):(S)-2,6-Diaminocaproic acid
- Melting point:215 °C (dec.) (lit.)
- Formula:H₂N(CH₂)₄CH(NH₂)CO₂H
- Molecular weight:146,19
- Flash point:Not applicable
- Form:Neat