Supelco® Organic Reference Standard, Ginsenoside Rg1
68317-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:22427-39-0
- Description:Ginsenoside Rg1
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00210293
- M-Clarity quality level:MQ100
- Synonym(s):(3β,6α,12β)-3,12-Dihydroxydammar-24-ene-6,20-diyl bis-β-ᴅ-glucopyranoside; Ginsenoside A2; Ginsenoside g1; Panaxoside A; Panaxoside Rg1; Sanchinoside C1; Sanchinoside Rg1
- Molecular weight:801,01
- GHS signal: GHS07