Organic reference standard, ethyl vanillin, analytical standard, Supelco®
Catalog # 75042-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:121-32-4
- Description:Ethyl vanillin, ≥98,5% (GC)
- Melting point:74...77 °C (lit.)
- Formula:C₉H₁₀O₃
- Description:Ethyl vanillin
- Form:Neat
Specifications
About this item
Ethyl vanillin is used as a reference standard in the quantitative determination of its presence in vanillin extract products using headspace-solid phase microextraction (HS-SPME) combined with gas chromatography coupled with mass spectrometry (GC–MS) and liquid chromatography coupled with mass spectrometry (LC-MS).