Phytochemical standard, avicularin, analytical standard, Supelco®
44006-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:572-30-5
- Description:Avicularin, ≥90,0% (HPLC)
- Environmentally Preferable:
- Formula:C₂₀H₁₈O₁₁
- Description:Avicularin
- Form:Neat