Order Entry
United Kingdom
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Aconitine
Catalog # 44787-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Aconitine
Catalog # 44787-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    302-27-2
  • Description:
    Aconitine
  • Environmentally Preferable:
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H300 + H330
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00082375
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Acetylbenzoylaconine
  • Formula:
    C₃₄H₄₇NO₁₁
  • Molecular weight:
    645,74
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06
Caution: P301 + P310 + P330 - P304 + P340 + P310