Order Entry
United Kingdom
ContactUsLinkComponent
Organic reference standard, Acesulfame K, for food analysis, ≥99,0%, Supelco®
Organic reference standard, Acesulfame K, for food analysis, ≥99,0%, Supelco®
Supplier:  Merck
Certificates
Organic reference standard, Acesulfame K, for food analysis, ≥99,0%, Supelco®
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    55589-62-3
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₄H₄KNO₄S
  • Description:
    Acesulfame K

Specifications

Related Information

Frequently Bought Together