Supelco® Cerilliant® Organic Reference Standard, Vitamin D2 solution
V-024-1ML
: Merck
:
- Concentration:1 mg/ml in ethanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:50-14-6
- Description:Vitamin D2 solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H319 - H373
- Shipping conditions:DICE
- MDL No.:MFCD00166988
- M-Clarity quality level:MQ300
- Synonym(s):Vitamin D2 solution
- Melting point:–144.0 °C
- Molecular weight:396,65
- Flash point:14,0 °C
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS08, GHS07
: P210 - P233 - P240 - P241 - P305 + P351 + P338 - P314