Supelco® Cerilliant® Organic Reference Standard, 1α,25-Dihydroxyvitamin D2 solution
Catalog # H-090-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:5 µg/ml in ethanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:60133-18-8
- Description:1α,25-Dihydroxyvitamin D2 solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H319
- Shipping conditions:DICE
- M-Clarity quality level:MQ300
- Molecular weight:428,65
- Flash point:14 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338