Supelco® PESTANAL® Organic Reference Standard, S-Hydroprene
Catalog # 46426-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:65733-18-8
- Description:S-Hydroprene
- Storage conditions:2...30 °C
- Hazard class description:H331 - H410
- Shipping conditions:STD
- MDL No.:MFCD03095706
- M-Clarity quality level:MQ100
- Synonym(s):Ethyl 3,7,11-trimethyl-2,4-dodecadienoate
- Molecular weight:266,42
- Flash point:>100 °C
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P261 - P271 - P273 - P304 + P340 + P311 - P391 - P403 + P233