Order Entry
United Kingdom
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Flumethrin
  46417-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Flumethrin
  46417-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    69770-45-2
  • Description:
    Flumethrin
  • Storage conditions:
    RT
  • Hazard class description:
    H301 + H311 + H331 - H315 - H317 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00867373
  • M-Clarity quality level:
    MQ200
  • Synonym(s):
    Cyano(4-fluoro-3-phenoxyphenyl)methyl 3-[2-chloro-2-(4-chlorophenyl)ethenyl]-2,2-dimethylcyclopropanecarboxylate
  • Molecular weight:
    510,38
  • Flash point:
    104 °C - Closed cup

 

 

  • GHS signal: GHS06
 : P261 - P280 - P301 + P310 - P302 + P352 + P312 - P304 + P340 + P311 - P305 + P351 + P338