Hema - rapid staining set, Gurr™ for hematology, for microscopy
Catalog # 351042L
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 KIT
- Seq:0075
- Environmentally Preferable:
Specifications
Specification Test Results
- Suitability for microscopy (blood smear):Passes test
- Nuclei:Red-violet
- Erythrocytes:Pink to brownish
- Eosinophilic granules:Red to red-brown
- Neutrophilic granules:Light violet
- Lymphocyte cytoplasm:Blue
Specification Test Results
About this item
:
:1