Pharmaceutical analytical standard, 6-Propyl-2-thiouracil-13C,15N2, reference material, Honeywell Fluka™
HONCR1502-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- Description:6-Propyl-2-thiouracil-¹³C,¹⁵N₂
- Environmentally Preferable:
- Formula:C₆¹³CH₁₀¹⁵N₂OS
- Storage temperature:Ambient
- Description:6-Propyl-2-thiouracil-¹³C,¹⁵N₂
- Freezing point:216...217 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results