Organic analytical standards, Stearic acid glycidylester-d5, neat, reference material, Honeywell Fluka™
HONCR1484-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1346598-19-3
- Description:Stearic acid glycidylester-d₅
- Formula:C₂₁H₃₅O₃D₅
- Storage temperature:Ambient
- Description:Stearic acid glycidylester-d₅
- Freezing point:52...53 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results