Pharmaceutical analytical standard, Caspofungin diacetate, reference material, Honeywell Fluka™
Catalog # HONCR1329-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:179463-17-3
- Description:Caspofungin diacetate
- Formula:C₅₂H₈₈N₁₀O₁₅ · 2C₂H₄O₂
- Storage temperature:–20 °C
- Description:Caspofungin diacetate
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results