Order Entry
United Kingdom
ContactUsLinkComponent
Pesticide reference standard, Clomeprop, neat, Honeywell Fluka™
Pesticide reference standard, Clomeprop, neat, Honeywell Fluka™
Catalog # HONCR1059-10MG
CAS Number:  
Pesticide reference standard, Clomeprop, neat, Honeywell Fluka™
Catalog # HONCR1059-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    84496-56-0
  • Description:
    Clomeprop
  • Formula:
    C₁₆H₁₅Cl₂NO₂
  • Storage temperature:
    2...8 °C
  • Description:
    Clomeprop
  • Freezing point:
    146...150 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results