Order Entry
United Kingdom
ContactUsLinkComponent
bisBenzimide H-33342 trihydrochloride trihydrate (Hoechst 33342)
Corrosive
Irritant
bisBenzimide H-33342 trihydrochloride trihydrate (Hoechst 33342)
Catalog # 40047.
Supplier:  Biotium
CAS Number:  
Corrosive
Irritant
bisBenzimide H-33342 trihydrochloride trihydrate (Hoechst 33342)
Catalog # 40047.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :615.99 g/mol (561.94 anhy)
EINECS:245-691-1
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
CAS Number:23491-52-3
Formula:C₂₇H₂₈N₆O·3HCl·3H₂O
MDL Number:MFCD00012679