Cell culture media, Dulbecco’s modified eagle’s medium (DMEM), high glucose
Supplier: Merck
Certificates
About this item
Dulbecco′s modified eagle′s medium (DMEM) is a modification of basal medium eagle (BME) that contains four-fold concentrations of the amino acids and vitamins.
This DMEM-High glucose medium is a 1X complete medium with sodium pyruvate added. It also differs from the original DMEM-High formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.
Frequently Bought Together