Supelco® Drug and metabolite standards, Quetiapine fumarate solution, (CRM)
Q-001-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:111974-72-2
- Description:Quetiapine fumarate solution
- Description:Quetiapine fumarate solution
- Pk Info:ampoule of 1 ml