Supelco® Drug and metabolite standards, Protriptyline hydrochloride solution, (CRM)
P-903-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1225-55-4
- Description:Protriptyline hydrochloride solution
- Environmentally Preferable:
- Description:Protriptyline hydrochloride solution
- Pk Info:ampoule of 1 ml