Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Supelco® Drug and metabolite standards, Lorcaserin-D4-maleate, (CRM)
Supelco® Drug and metabolite standards, Lorcaserin-D4-maleate, (CRM)
Supelco® Drug and metabolite standards, Lorcaserin-D4-maleate, (CRM)
Catalog # SIALB130786-10MG
Supplier:
Merck
CAS Number:
Expand image
Supelco® Drug and metabolite standards, Lorcaserin-D4-maleate, (CRM)
Catalog # SIALB130786-10MG
Supplier:
Merck
CAS Number:
Specifications
Pk:
10 mg
Description:
Lorcaserin-D4-maleate
Specifications
About this item
Product Details
Product Details