Order Entry
United Kingdom
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, Dehydroepiandrosterone-D5 (DHEA-D5) (2,2,3,4,4-D5) solution,, (CRM)
  D-064-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, Dehydroepiandrosterone-D5 (DHEA-D5) (2,2,3,4,4-D5) solution,, (CRM)
  D-064-1ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    97453-25-3
  • Description:
    Dehydroepiandrosterone-D5 (DHEA-D5) (2,2,3,4,4-D5) solution,
  • Description:
    Dehydroepiandrosterone-D5 (DHEA-D5) (2,2,3,4,4-D5) solution,
  • Pk Info:
    ampoule of 1 ml