Order Entry
United Kingdom
ContactUsLinkComponent
Supelco® Drug and metabolite standards, cis-Tramadol hydrochloride solution, (CRM)
Supelco® Drug and metabolite standards, cis-Tramadol hydrochloride solution, (CRM)
Catalog # T-027-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, cis-Tramadol hydrochloride solution, (CRM)
Catalog # T-027-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    22204-88-2
  • Description:
    cis-Tramadol hydrochloride solution
  • Description:
    cis-Tramadol hydrochloride solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item