Order Entry
United Kingdom
ContactUsLinkComponent
Drug and metabolite standards, 17alpha-Hydroxypregnenolone-13C2,D2 solution, (CRM), Supelco®
Drug and metabolite standards, 17alpha-Hydroxypregnenolone-13C2,D2 solution, (CRM), Supelco®
Catalog # H-112-1ML
Supplier:  Merck
CAS Number:  
Drug and metabolite standards, 17alpha-Hydroxypregnenolone-13C2,D2 solution, (CRM), Supelco®
Catalog # H-112-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    17alpha-Hydroxypregnenolone-13C2
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item