Supelco® Organic Reference Standard, trans-9-Octadecenoic acid methyl ester
Catalog # CRM46903
Supplier: Merck
CAS Number:
Specifications
- Concentration:10 mg/ml in heptane
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1937-62-8
- Description:trans-9-Octadecenoic acid methyl ester
- Storage conditions:–10...–25 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:DICE
- MDL No.:MFCD00066521
- M-Clarity quality level:MQ100
- Synonym(s):Methyl elaidate; Elaidic acid methyl ester; Methyl trans-9-octadecenoate
- Formula:CH₃(CH₂)₇CH=CH(CH₂)₇COOCH₃
- Molecular weight:296,49
- Density:0,871 g/ml at 20 °C (lit.)
- Flash point:–3,99 °C – Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331