Supelco® Organic Reference Standard, ʟ-Ornithine monohydrochloride
57197-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:3184-13-2
- Description:ʟ-Ornithine monohydrochloride
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00064562
- M-Clarity quality level:MQ100
- Synonym(s):(S)-2,5-Diaminopentanoic acid monohydrochloride
- Molecular weight:168,62