Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Neutralising solution, NLL
NLL neutraliser, Plastic container
Catalog # LANC41010014_
Neutralising solution, NLL
Catalog # LANC41010014_
Neutralising solution, NLL
NLL neutraliser, Plastic container
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
NLL neutraliser is formulated for washing of laboratory glassware.
  • Phosphate-free
  • Non foaming
NLL neutraliser is used for borosilicate glass, soda-lime glass, fused quartz glass, polymeric material, porcelain and more.
Specifications
  • Description:
    NLL neutraliser
  • Packed:
    Plastic container